Search Thermo Fisher Scientific
Catalog Number | Holds | Positions | For Use With (Equipment) |
---|---|---|---|
FKS11369321 | Boxes (2 in.) | 16 Boxes/Rack | Upright Freezers (17.3, 23 cu.ft.) |
398326 | Boxes (2 in.) | 16 Boxes/Rack | Upright Freezers (17.3, 23 cu.ft.) |
398331 | Boxes (2 in.) | 16 Boxes/Rack | Upright Freezers (17.3, 23 cu.ft.) |
Maximize the storage space in your ultra-low temperature freezer with these Thermo Scientific™ Racks for TSX/TDE 5 shelf Series Freezers as well as well as previous generation Revco™ ExF, DxF and HERAfeeze™ HFU B Freezers. Includes sliding drawer racks, adjustable side access box racks and microplate racks in standard or deepwell formats.
Thermo Scientific™ freezer racking and tubing solutions offer a convenient, streamlined process to maximize sample storage in your laboratory. Our racking solutions are designed to hold tubes ranging from 200μL up to 5.0mL, offering maximized sample storage capacity.