SP Scienceware™ Micro Spoon and Spatula Sampling Set
SP Scienceware™ Micro Spoon and Spatula Sampling Set
Actual product may vary
Fisherbrand
SP Scienceware™ Micro Spoon and Spatula Sampling Set
Includes six different, small-size, double-ended tools and tweezers for accurate handling of small quantities. Bel Art™ SP Scienceware™ Micro Spoon and Spatula Sampling Set are polished stainless steel for long life and easy cleaning and can be steam autoclaved at 121ºC (250ºF)
Have Questions?
Catalog NumberMaterial
FKS11352204Stainless Steel
Catalog number FKS11352204
Price (MYR)
1,364.00
EA
Add to cart
Material:
Stainless Steel
Price (MYR)
1,364.00
EA
Add to cart
Includes six different, small-size, double-ended tools and tweezers for accurate handling of small quantities. Bel Art™ SP Scienceware™ Micro Spoon and Spatula Sampling Set are polished stainless steel for long life and easy cleaning and can be steam autoclaved at 121ºC (250ºF)
  • Each tool has two different end shapes, doubling the selection of implements to better match user needs
  • Tool ends include 2 small spoons for transferring and collecting both dry and liquid material
  • Eight flat spatulas of various widths and lengths excellent for mixing, scraping and spreading; Six of these have a square end and two are rounded
  • Two V-shaped spatulas of different sizes can be used for lifting or spill-free carrying of material
  • The blunt-tip tweezers have serrated inside surfaces for secure sample handling and grooved handles for better gripping
  • All tools are polished stainless steel for long life and easy cleaning and are steam autoclavable at 121°C (250°F)
  • A carrying case protects and organizes all tools when not in use
Specifications
Product TypeMicro Spoon and Spatula Sampling Set
MaterialStainless Steel
AutoclavableAutoclavable
Includes6 small size double-ended tools, set of tweezers, 2 small spoons, 6 square end flat spatulas, 2 round end flat spatulas, 2 v-shaped spatulas, blunt tip tweezers, carrying case
DisposableReusable
For Use With (Application)Transferring and collecting both dry and liquid material
Unit SizeEA