Poxygrid™ Petri Dish Dispensing Racks
Poxygrid™ Petri Dish Dispensing Racks
Actual product may vary
Poxygrid™ Petri Dish Dispensing Racks
Poxygrid™ Petri Dish Dispensing Racks
Fisherbrand
Poxygrid™ Petri Dish Dispensing Racks
Holds 11 to 33 plastic petri dishes. Bel-Art™ Poxygrid™ Petri Dish Dispensing Racks are durable and dispense dishes one at a time.
Have Questions?
Catalog NumberPositionsDimensions (L x W x H)
FKS11352194115.75 x 4.25 x 11 in.
Catalog number FKS11352194
Price (MYR)
827.00
EA
Add to cart
Positions:
11
Dimensions (L x W x H):
5.75 x 4.25 x 11 in.
Price (MYR)
827.00
EA
Add to cart
Holds 11 to 33 plastic petri dishes. Bel Art™ SP Scienceware™ Poxygrid™ Petri Dish Dispensing Racks are durable and dispense dishes one at a time.
  • Durable epoxy-coated, steel wire construction
  • Dishes loaded at top and dispensed at bottom
  • Each channel holds up to eleven 100 x 20 mm plastic Petri dishes
  • Tilts toward rear for convenient presentation of dishes
  • Not suitable for glass Petri dishes
Specifications
IncludesRack Only
Product TypePetri Dish Rack
Array1 x 1
AutoclavableNot Autoclavable
ColorBlue
HoldsPetri Dishes
MaterialSteel Wire
Positions11
ShapeCylindrical
StackableNo
SterilityNon-sterile
For Use With (Equipment)10 x 2cm Petri dishes
Dimensions (L x W x H)5.75 x 4.25 x 11 in.
Length (Metric)14.6 cm
Length (English)5.75 in.
Width (Metric)10.7 cm
Width (English)4.25 in.
Height (Metric)28 cm
Height (English)11 in.
Diameter (Metric) Inner100 mm
Unit SizeEA