Nunc™ Conical Sterile Polypropylene Centrifuge Tubes
Nunc™ Conical Sterile Polypropylene Centrifuge Tubes
Actual product may vary
Nunc™ Conical Sterile Polypropylene Centrifuge Tubes
These premium, high-quality conical tubes are environmentally friendly, and allow for increased traceability with the largest writing area on the market.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)PackagingQuantity
FKS1134720150 mLBulk25 Pk.
FIS12-565-26815 mLBulk50 Pk.
FIS12-565-27050 mLBulk25 Pk.
Catalog number FKS11347201
Price (MYR)
3,359.00
PK
Add to cart
Capacity (Metric):
50 mL
Packaging:
Bulk
Quantity:
25 Pk.
Price (MYR)
3,359.00
PK
Add to cart
Thermo Scientific™ Nunc Conical Sterile Polypropylene Centrifuge Tubes are premium, high-quality conical tubes that are environmentally friendly, offer the highest cleanliness with a recyclable plastic rack and allow for increased traceability with the largest writing area on the market. Nunc Labware Products are made from high-purity resins and molded using our state-of-the-art processes. Plastic labware is a safer alternative to glass without sacrificing accuracy.
Features
  • Nonpyrogenic: Endotoxin tested (0.5 EU/mL)
  • Non-Cytotoxic
  • Biologically inert: USP biological reactivity class VI
  • High level of sterility: SAL 10-6
  • RNase/DNase-free
  • Leak-proof to protect your samples
  • Made from high-purity resins and molded using state-of-the-art processes; safe alternative to glass without sacrificing accuracy
  • Racked product includes environmentally friendly recyclable plastic rack that reduces landfill waste compared to the traditional foam packaging
  • Plug sealed cap
  • Tube, cap and rack made of virgin resin
Warranty: 90 days
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Closure ColorBlue
ColorClear
Capacity (Metric)50 mL
DisposableSingle-use
MaterialPolypropylene
No. per Case500/Cs.
No. per Pack25
PackagingBulk
Quantity25 Pk.
SterilitySterile
Green FeaturesSustainable packaging
RNase FreeYes
Dnase FreeYes
AutoclavableNot Autoclavable
ShapeCylindrical
Length (Metric)98.6 mm
Bottom ShapeConical
Closure TypeScrew Cap
Closure IncludedYes
Writing AreaYes
GraduatedYes
Product TypeCentrifuge Tube
Max. RCF17,000 x g
Unit SizePK