Clear Glass Ointment Jars with Black Urea Screw Caps
Clear Glass Ointment Jars with Black Urea Screw Caps
Actual product may vary
Fisherbrand
Clear Glass Ointment Jars with Black Urea Screw Caps
Manufactured from soda lime glass. Azpack™ Clear Glass Ointment Jars with Black Urea Screw Caps can be used to store creams, powders, and liquids.
Have Questions?
Catalog NumberCapacity (Metric)Closure Size
FKS11337034250 mL38/R3
Catalog number FKS11337034
Price (MYR)
434.00
PK
Add to cart
Capacity (Metric):
250 mL
Closure Size:
38/R3
Price (MYR)
434.00
PK
Add to cart
  • Soda lime glass bottle
  • Extra wide neck
  • Includes wood pulp-faced PVDC-lined screw cap
Specifications
Capacity (Metric)250 mL
ColorClear
MaterialSoda Lime Glass
Quantity10/PK
Closure ColorBlack
Closure IncludedYes
MouthExtra Wide
Height (Metric)73.8 mm
Closure LinerWood Pulp-faced PVDC
Closure MaterialUrea
Closure Size38/R3
Diameter (Metric)87 mm
Closure TypeScrew Cap
Unit SizePK