Glass Sample Vials
Glass Sample Vials
Actual product may vary
Fisherbrand
Glass Sample Vials
Sample and contain pathological materials. Azpack™ Glass Sample Vials include synthetic rubber-lined aluminum caps that should be removed before autoclaving.
Have Questions?
Catalog NumberColor
FKS11328053Clear
Catalog number FKS11328053
Price (MYR)
4,016.00
PK
Add to cart
Color:
Clear
Price (MYR)
4,016.00
PK
Add to cart
  • J669 Universal Vial
  • Type 2 soda lime silica glass construction
  • Includes aluminum cap with black synthetic rubber liner
  • Vials, caps, and liners are all fully autoclavable
  • Remove caps/liners from the vial during autoclaving to prevent deterioration after repeated autoclaving
Specifications
Material (Liner)Synthetic Rubber
ColorClear
Diameter (Metric) Outer29.50 mm
MaterialSoda Lime Glass
Volume (Metric)28 mL
For Use With (Application)Pathology
Closure MaterialAluminum
AutoclavableAutoclavable
Closure TypeScrew Cap
Height (Metric)83.50 mm
Quantity144 Pk.
Closure SizeR3/28
Unit SizePK