Space Saver Vacuum Desiccator
Space Saver Vacuum Desiccator
Actual product may vary
Space Saver Vacuum Desiccator
Degas liquids quickly and easily in the laboratory or classroom with these small plastic vacuum desiccators.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorHeight (English)Height (Metric)Material
FKS11326984White10.25 in.26 cmPolypropylene, Polycarbonate
FIS08-594-15CWhite12.12 in.31 cmPolypropylene, Polycarbonate
FKS10236044Clear10.25 in.26 cmPolycarbonate
Catalog number FKS11326984
Price (MYR)
1,985.00
EA
Add to cart
Color:
White
Height (English):
10.25 in.
Height (Metric):
26 cm
Material:
Polypropylene, Polycarbonate
Price (MYR)
1,985.00
EA
Add to cart
Degas liquids quickly and easily in the laboratory or classroom with these small plastic vacuum desiccators.
  • Strong, shatterproof units provide dry storage and protection for sensitive products
  • Lighter and less costly than similarly sized glass models; do not chip like glass
  • Hold a full vacuum (29.9 in. Hg) at room temperature for 24 hours
  • Tapered polypropylene stopcock with PTFE plug accepts tubing with 0.25 in. to 0.375 in. I.D. and allows a controlled entrance of air, preventing unnecessary turbulence
  • Neoprene O-ring assures a tight seal down to 0°C without the need for grease (ring will not be harmed if grease is used)
  • Flat Dome clear polycarbonate top with large ribbed knob maximizes interior clearances and provides an average of 13% greater interior volume over standard hemispherical domes
  • All models include a 0.125 in. (0.32 cm) thick perforated plate; disposable or reusable desiccant can be placed on or under this plate
  • Clear polycarbonate bottom models offer increased viewing visibility from more angles
  • Models with white polypropylene bottoms can support incandescent crucibles on Minerit HD High Heat Ceramic Desiccator Plates (available separately, F42038 series)
  • Models with clear polycarbonate bottoms offer increased visibility from more angles
Specifications
ColorWhite
MaterialPolypropylene, Polycarbonate
Product TypeVacuum Desiccator
IncludesUpper and lower sections, neoprene O-Ring, stopcock, 0.32cm thickness polypropylene perforated sample plate, tapered polypropylene stopcock with PTFE plug and red handle, flat dome clear polycarbonate top with large ribbed knob
Height (Metric)26 cm
Height (English)10.25 in.
Diameter (English) Inner7.75 in.
Diameter (English) Outer Flange9.06 in.
Diameter (English) Plate7.4 in.
Diameter (Metric) Outer Flange23 cm
Diameter (Metric) Plate19 cm
Diameter (Metric) Inner19.7 cm
Diameter (Metric) Outer23.3 cm
Unit SizeEA