Whatman™ Cellulose Soxhlet Extraction Thimbles, Single 1mm Wall
Whatman™ Cellulose Soxhlet Extraction Thimbles, Single 1mm Wall
Actual product may vary
Whatman™ Cellulose Soxhlet Extraction Thimbles, Single 1mm Wall
Whatman™ Cellulose Soxhlet Extraction Thimbles, Single 1mm Wall
Whatman™ Cellulose Soxhlet Extraction Thimbles, Single 1mm Wall
Manufactured from high-quality alpha cellulose cotton linter for high-performance use in automated Soxhlet extractor systems. Whatman™ Cellulose Soxhlet Extraction Thimbles are intended for applications including use in air and waste gas analysis.
Have Questions?
Catalog NumberDiameter (Metric) InsideLength (Metric)
FKS1132070425 mm80 mm
Catalog number FKS11320704
Price (MYR)
1,613.00
PK
Add to cart
Diameter (Metric) Inside:
25 mm
Length (Metric):
80 mm
Price (MYR)
1,613.00
PK
Add to cart
Manufactured from high-quality alpha cellulose cotton linter for high-performance use in automated Soxhlet extractor systems. Whatman™ Cellulose Soxhlet Extraction Thimbles are intended for applications including use in air and waste gas analysis.
  • Used in a wide variety of solid-liquid extraction studies, including lipid and pesticide analyses in food and soil samples
  • Suitable for temperatures up to 120°C (248°F)
  • Excellent mechanical strength and retention
  • Wall thickness of standard thimbles is 1mm to support application need
Specifications
Quantity25 Pk.
DescriptionGE’s Whatman cellulose thimbles are manufactured from high-quality alpha cellulose cotton linter for high-performance use in automated Soxhlet extractor systems. Used in air and waste gas analysis.
Diameter (Metric) Inside25 mm
MaterialCellulose
Wall Thickness1 mm
For Use With (Application)Air and waste gas analysis
For Use With (Equipment)Soxhlet extractor
Length (Metric)80 mm
Max. Temperature (Metric)120°C
Particle Retention10μm
Product TypeExtraction Thimble
Unit SizePK