Check Order Status
Aluminum Crimp Seals
Actual product may vary
Aluminum Crimp Seals
Seals freeze drying and injection vials. Azpack™Aluminum Crimp Seals prevent leakage of hazardous and biological products.
Have Questions?
Catalog Number
Diameter (Metric) Outer
Volume (Metric)
FKS11318133
20 mm
5, 10, 20 mL
Catalog number
FKS11318133
Price (MYR)
931.00
PK
Add to cart
Diameter (Metric) Outer:
20 mm
Volume (Metric):
5, 10, 20 mL
Price (MYR)
931.00
PK
Add to cart
For use with clear neutral glass vials
Aluminum
500/Pack
Specifications
Color
Silver
Diameter (Metric) Outer
20 mm
Material
Aluminum
Volume (Metric)
5, 10, 20 mL
Closure Type
Crimp Cap