Whatman™ Quantitative Filter Paper: Grade 589/1 Circles
Whatman™ Quantitative Filter Paper: Grade 589/1 Circles
Actual product may vary
Fisherbrand
Whatman™ Quantitative Filter Paper: Grade 589/1 Circles
Designed for fast filtration of coarse precipitate. Whatman™ Grade 589/1 Quantitatve FIlter Paper Circles support tests such as cement fineness measurement in a standard Blaine test or determination of the ash content in foodstuffs.
Have Questions?
Catalog NumberDiameter (Metric)
FKS11315194150 mm
Catalog number FKS11315194
Price (MYR)
1,144.00
PK
Add to cart
Diameter (Metric):
150 mm
Price (MYR)
1,144.00
PK
Add to cart
Designed for fast filtration of coarse precipitate. Whatman™ Grade 589/1 Quantitatve FIlter Paper Circles support tests such as cement fineness measurement in a standard Blaine test or determination of the ash content in foodstuffs.
Ashless grade are very pure filters suitable for a wide range of critical analytical filtration procedures. Whatman™ quantitative filter papers are designed for gravimetric analysis and the preparation of samples for instrumental analysis.
  • For coarse precipitate retention
  • Herzberg filtration speed of 50 s
  • Black Ribbon Filter for filtration of coarse precipitates
  • Ashless with very high flow rate
  • High purity (< 0.01% ash)
Specifications
Color CodeBlack
DescriptionGrade 589/1 Filter for Coarse Precipitate Retention, 150 mm circle (100 pcs)
Filter TypeAshless
FormatDisc
MaterialCotton Linters
Particle Retention12 to 15 μm
Filtration Speed25 s
Flow RateHigh
For Use With (Application)Designed for gravimetric analysis and the preparation of samples for instrumental analysis.For the filtration of coarse precipitates.For many quantitative standard methods, especially for gravimetric applications (e.g., determination of the ash content
GradeGrade 589/1
Thickness (Metric)190 μm
TypeQuantitative Filter Paper
Diameter (Metric)150 mm
Unit SizePK