Lab-Aire™ II Glassware Drying Racks
Lab-Aire™ II Glassware Drying Racks
Actual product may vary
Lab-Aire™ II Glassware Drying Racks
Lab-Aire™ II Glassware Drying Racks
Lab-Aire™ II Glassware Drying Racks
Glassware and plasticware drying racks for walls or benchtops
Have Questions?
Catalog NumberPositionsProduct TypeHeight (English)Depth (English)
FKS1130275457Wall-mount Rack (3-tier)23.4 in.5 in.
Catalog number FKS11302754
Price (MYR)
3,703.00
EA
Add to cart
Positions:
57
Product Type:
Wall-mount Rack (3-tier)
Height (English):
23.4 in.
Depth (English):
5 in.
Price (MYR)
3,703.00
EA
Add to cart
Glassware and plasticware drying racks for walls or benchtops
  • Dry and store delicate labware including beakers, flasks, bottles, test tubes and cylinders on these space-saving drying racks
  • Single-sided benchtop models fit flat against a wall
  • Double-sided benchtop models provide double the capacity (up to 114 pegs) with a slightly larger footprint
  • Three sizes and lengths of V-channel pegs can be repositioned to hold a variety of labware
  • Rugged ABS plastic body and pegs wipe clean and won't corrode
  • Wall-mount models have mounting points that fit standard pegboards
  • Pegs available separately to reconfigure rack for specific drying requirements
Specifications
IncludesPegs
Product TypeWall-mount Rack (3-tier)
AutoclavableNot Autoclavable
ColorCream, Teal
HoldsLab Glassware
MaterialABS
Positions57
ShapeRectangular
ShelvesNone
StackableNo
SterilityNon-sterile
For Use With (Equipment)Lab Glassware
Size375 x 239 mm
Length (Metric)239 mm
Width (Metric)37.5 cm
Width (English)14.75 in.
Height (Metric)59.4 cm
Height (English)23.4 in.
Depth (English)5 in.
Depth (Metric)12.7 cm
Unit SizeEA