Broken Glass Disposal Box
Broken Glass Disposal Box
Actual product may vary
Fisherbrand
Broken Glass Disposal Box
Ruggedly constructed, puncture-resistant glass disposal receptacles safely collect broken and discarded laboratory glass. Boxes are clearly labeled, and highly visible.
Have Questions?
Catalog NumberCapacity (English)
FKS113022042.3 gal.
Catalog number FKS11302204
Price (MYR)
1,313.00
PK
Add to cart
Capacity (English):
2.3 gal.
Price (MYR)
1,313.00
PK
Add to cart
Ruggedly constructed, puncture-resistant glass disposal receptacles safely collect broken and discarded laboratory glass. Boxes are clearly labeled, and highly visible.
  • Sturdy corrugated cardboard receptacle is supplied with a 2 mil thick polyethylene bag to contain broken glass fragments and spilled liquids
  • Printed with eye-catching graphics in four languages (English, German, French, Spanish)
  • Packaged flat to save space and shipping costs and is quickly assembled
  • Does not qualify as a DOT container
  • Safe puncture resistant receptacles for broken glassware
  • Floor model holds 40 lb. (18 kg) of waste
Specifications
Capacity (Metric)10 L
Capacity (English)2.3 gal.
ColorGreen
Dimensions (L x W x H)12 x 12 x 27 in. (30 x 30 x 69 cm)
Height (Metric)69 cm
Length (Metric)30 cm
Width (Metric)30 cm
AutoclavableNot Autoclavable
Width (English)12 in.
Length (English)12 in.
Height (English)27 in.
MaterialCorrugated Cardboard
Product TypeSafety Container
Unit SizePK