Blaubrand™ Class A Borosilicate Glass Volumetric Flasks with Glass Stopper
Actual product may vary
Blaubrand™ Class A Borosilicate Glass Volumetric Flasks with Glass Stopper
Constructed of Borosilicate Glass and complies with DIN EN ISO 1042 standards. BRAND™ Blaubrand™ Class A Volumetric Flask with Glass Stopper is available in both narrow neck and wide neck options in a variety of sizes. DE-M marking on neck provides quick and accurate volume check.
Have Questions?
Catalog Number
Capacity (Metric)
Closure Size
Neck Style
FKS10759852
1000 mL
24/29
Narrow
Catalog number FKS10759852
Price (MYR)
550.00
EA
Add to cart
Capacity (Metric):
1000 mL
Closure Size:
24/29
Neck Style:
Narrow
Price (MYR)
550.00
EA
Add to cart
Class A Volumetric Flask
Constructed of Borosilicate Glass
Complies with DIN EN ISO 1042 standards
DAkkS calibration certificate available upon request
Calibrated to contain (TC, In)
Narrow neck and wide neck options available in a variety of sizes
DE-M marking provides quick and accurate volume check