Orion™ pH Buffer Individual Use Pouches
Orion™ pH Buffer Individual Use Pouches
Actual product may vary
Orion™ pH Buffer Individual Use Pouches
Ensure fresh, ready-to-use standards for pH calibration with these pH Buffer individual use pouches.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantitypH
FKS1075686125 x 15 mL4.01
FKS1024249125 x 15 mL7
FKS1047044325 x 15 mL10.01
Catalog number FKS10756861
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 x 15 mL
pH:
4.01
Ensure fresh, ready-to-use standards for pH calibration with Thermo Scientific™ Orion™ pH Buffer Individual Use Pouches. Buffers are color coded for ease of use. All Orion pH buffers are traceable to NIST™ standard reference materials.
  • Color coded: pH 4.01 buffer is pink, pH 7.00 buffer is yellow, and pH 10.01 buffer is blue
  • Packaged with buffer/temperature tables for accurate calibrations and NIST traceability
  • Manufactured under ISO 9000 quality standards
  • Pouches are 15mL each and are available in 10 packs or 25 packs
Note:
The following part numbers have been changed due to a recent system upgrade.
Old Part Number: 910410
New Part Number: 910410-WA
Old Part Number: 911025
New Part Number: 911025-WA
Please use the new part number when placing an order or requesting a quote. The old and new part numbers are interchangeable on Certificate of Analysis and Material Safety Data Sheet (MSDS) files.
Warranty: Out of box
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Chemical Name or MaterialpH Buffer, individual use pouches
Boiling Point100°C
ColorLight Red
DescriptionpH 4.01 Buffer
For Use With (Application)pH calibration
For Use With (Equipment)pH Meter and Electrode, single-use
pH4.01
Physical FormLiquid
Quantity25 x 15 mL
Solution TypeBuffer
FormulationWater, Potassium Hydroxide, Amaranth Red No. 27, Potassium Hydrogen Phthalate
PackagingPouch
Traceability to NISTYes
Solubility InformationSoluble
Unit SizePK