Borosilicate Glass Class AS Burette with Teflon Tap
Borosilicate Glass Class AS Burette with Teflon Tap
Actual product may vary
Fisherbrand
Borosilicate Glass Class AS Burette with Teflon Tap
Complies with BS-IS 385 standards. Fisherbrand™ Borosilicate Glass Class AS Burette with Teflon Tap includes single bore with interchangeable PTFE stopcock. Blue ceramic graduations make volume assessment easy.
Have Questions?
Catalog NumberCapacity (Metric)
FKS1073808150 mL
Catalog number FKS10738081
Price (MYR)
481.00
EA
Add to cart
Capacity (Metric):
50 mL
Price (MYR)
481.00
EA
Add to cart
  • Constructed of borosilicate glass
  • Single bore with interchangeable PTFE stopcock
  • Blue ceramic graduations
  • Class AS, BS-ISO 385 Compliance
Specifications
Capacity (Metric)50 mL
MaterialBorosilicate Glass
Graduations0.1 mL
IncludesSingle Bore, Interchangeable Stopcock
ScaleBlue
Stopcock MaterialPTFE
Tolerance±0.05 mL
TypeBuret
ClassClass AS
Unit SizeEA