Dual Tube Handheld UV Lamp
Dual Tube Handheld UV Lamp
Actual product may vary
Fisherbrand
Dual Tube Handheld UV Lamp
Select from longwave, midrange, shortwave or combination longwave and shortwave in the same lamp. UVP Ultra Violet Product™ Handheld UV Lamp uses two separate UV tubes for greater intensity than split-tube 6 watt models.
Have Questions?
Catalog number FKS10712467
Price (MYR)
6,665.00
EA
Add to cart
Price (MYR)
6,665.00
EA
Add to cart
  • Lightweight, A/C powered, and portable
  • 6 watt portable lamp
  • Wavelengths: 365nm longwave and 254nm shortwave.
  • Two separate tubes. Durable plastic construction
  • Lamp cord: 2.4m
  • Dimensions: 378 x 81 x 64mm
  • Weight: .09kg
  • Use with J-129 stand for hands-free operation.
Specifications
Voltage230 V
Wattage6 W
Wavelength365/254 nm
Dimensions (L x W x H)378 x 81 x 64 mm
Electrical Requirements6 W, 230 V, 50/60 Hz
Hertz50/60 Hz
Includes2 separate tubes
Lighting Intensity1350/1200 μWcm2
MaterialPlastic
ModelUVGL-58
Packaging Quantity1 Ea.
Product TypePortable UV Lamp
Unit SizeEA