Glass Microfiber Filters
Glass Microfiber Filters
Actual product may vary
Fisherbrand
Glass Microfiber Filters
Binder free glass microfiber filters are suitable for analytical and gravimetric analyses. They are also used as prefilter.
Have Questions?
Catalog NumberDiameter (Metric)ResistivityThickness (Metric)
FKS107070814.7 cm360.02 cm
Catalog number FKS10707081
Price (MYR)
680.00
PK
Add to cart
Diameter (Metric):
4.7 cm
Resistivity:
36
Thickness (Metric):
0.02 cm
Price (MYR)
680.00
PK
Add to cart
Binder free glass microfibre filters are made of 100 % borosilicate, are highly pure and extremely white. They continue to perform over long periods unlike cellulose filters whose rate of filtration drops off rapidly as the particulate load increases. They are ideal for critical analyses in which impurities can be washed out of the filter. Moreover, binder free glass microfibre filters are temperature resistant up to 500 or 550°C (grade MG 550-HA ).
They are available in different formats as in rolls, discs and sheets.
Specifications
Color (Overall)White
Filtration Time40 sec.
GradeMGA
Particle Retention1.6 μm
TypeFilter
FormatFilter Disc
For Use With (Application)Ideal for critical analyses in which impurities can be washed out of the filter
MaterialBorosilicate Glass
PurityHigh
Resistivity36
Temperature (Metric) Resistant500°C or 550°C
Thickness (Metric)0.02 cm
Diameter (Metric)4.7 cm
Weight (Metric)52
Unit SizePK