pH Electrode Storage and Cleaning Solutions
pH Electrode Storage and Cleaning Solutions
Actual product may vary
pH Electrode Storage and Cleaning Solutions
Keep your pH probes clean and properly stored for longevity
Have Questions?
Catalog NumberQuantity
FKS106601661 Each
Catalog number FKS10660166
Price (MYR)
-
Quantity:
1 Each
Keep your pH probes clean and properly stored for longevity
  • Remove grease and oil deposits from your electrodes with our cleaning solution.
  • Cleaning solution contains pepsin.
  • After cleaning, keep your electrodes in proper working condition with our storage solution.
Specifications
Boiling Point>100°C
Capacity (English)16.9 oz.
Capacity (Metric)500 mL
ColorClear
CompositionWater, potassium chloride, potassium dihydrogen phosphate, disodium hydrogen phoshpate
ConductivityNot tested
DescriptionElectrode Storage Solution; 500 mL
For Use With (Equipment)pH Electrode
TypeStorage Solution
Filling Solution TypepH Electrode Storage
For Use With (Application)pH testing
IncludesElectrode Storage Solution; 500 mL
NIST TraceableNo
OdorOdorless
Optical DensityNot tested
Quantity1 Each
Recommended StorageRoom Temperature
Shelf Life12 Months
SolubilityWater Soluble
Water SolubleYes
Dimensions (L x W x H)3 x 3 x 6.25 in.
Length (English)3 in.
Length (Metric)7.62 cm
Width (English)3 in.
Width (Metric)7.62 cm
Height (English)6.25 in.
Height (Metric)15.9 cm
Unit SizeEA