DURAN™ Woulff Bottle, 3 standard ground necks
DURAN™ Woulff Bottle, 3 standard ground necks
Actual product may vary
Fisherbrand
DURAN™ Woulff Bottle, 3 standard ground necks
The Woulff bottle is used in vacuum apparatus and serves to collect, clean and/or dry gases.
Have Questions?
Catalog NumberCapacity (Metric)
FKS10614501500 mL
Catalog number FKS10614501
Price (MYR)
1,576.00
EA
Add to cart
Capacity (Metric):
500 mL
Price (MYR)
1,576.00
EA
Add to cart
Woulff''s bottles are part of vacuum apparatus, in which they are used on the one hand as collecting vessels for gas generation or for cleaning and/or drying gases. As part of a vacuum apparatus, Woulff''s bottles are suitable for vacuum applications and often also used as an intermediate vessel to prevent spontaneous drop-off of the vacuum and the associated liquid suction back into the vacuum pump.
  • Tight closure due to standard ground joint
  • Vacuum resistant due to the wall thickness and geometry
  • Three standard ground joints for the import or export of gases
  • Optimum safety for working under pressure or vacuum
  • Fired-on, highly durable white print
Specifications
Capacity (Metric)500 mL
For Use With (Application)Collecting and dissolving gases in liquids; Production of gases; Safe working under vacuum; In series: Purification of a gas
MaterialBorosilicate Glass (3.3)
No. per Pack1/Pk.
AutoclavableAutoclavable
Diameter (Metric)87 mm
Unit SizeEA