PTFE Flat-Form Evaporating Dish with Spout
PTFE Flat-Form Evaporating Dish with Spout
Actual product may vary
Fisherbrand
PTFE Flat-Form Evaporating Dish with Spout
Includes spout for easy pouring. The PTFE flat-form evaporating dish with spout is available in a variety of sizes.
Have Questions?
Catalog NumberCapacity (Metric)Diameter (Metric)Height (Metric)
FKS1058239250 mL82 mm15 mm
Catalog number FKS10582392
Price (MYR)
193.00
EA
Add to cart
Capacity (Metric):
50 mL
Diameter (Metric):
82 mm
Height (Metric):
15 mm
Price (MYR)
193.00
EA
Add to cart
  • Isostatically molded in pure PTFE
  • Chemically resistant and inert
  • Super smooth internal finish
  • Usable to 280°C
  • Molded spout for easy pouring
Specifications
Capacity (English)1.66 fl oz.
Bottom ShapeFlat
Capacity (Metric)50 mL
MaterialPTFE
ShapeRound
TypeEvaporating Dish
IncludesSpout
Interior FinishSmooth
Temperature Range (Metric)-200°C to +280°C
Height (Metric)15 mm
Diameter (Metric)82 mm
Unit SizeEA