Spray Bottle
Spray Bottle
Actual product may vary
Fisherbrand
Spray Bottle
Dispense and spray fluids efficiently. Kartell™ Spray Bottle is ideal for dispensing a wide range of fluids including cleaners, cosmetics, and chemical specialties. The bottle features an atomizer with an adjustment jet.
Have Questions?
Catalog NumberCapacity (Metric)
FKS105796621000 mL
Catalog number FKS10579662
Price (MYR)
50.00
EA
Add to cart
Capacity (Metric):
1000 mL
Price (MYR)
50.00
EA
Add to cart
Dispense and spray fluids efficiently. Kartell™ Spray Bottle is ideal for dispensing a wide range of fluids including cleaners, cosmetics, and chemical specialties. The bottle features an atomizer with an adjustment jet.
  • Atomizers with adjustment
  • HDPE
  • Suitable for foodstuff
Image is exemplary - spray head might look differently
Specifications
Capacity (Metric)1000 mL
Diameter (Metric) Outer Neck90mm
Diameter (Metric) Inner22.5 mm
MaterialPlastic
Liner MaterialHDPE
Height (Metric)271 mm
Unit SizeEA