Aerosol Analysis Monitors, 37 mm
Aerosol Analysis Monitors, 37 mm
Actual product may vary
Fisherbrand
Aerosol Analysis Monitors, 37 mm
Millipore's 37 mm Aerosol Analysis Monitors are transparent, disposable, plastic filter holders for use with Type MF (mixed cellulose esters) Millipore filters. A thin cellulose support seals the filter between monitor sections to distribute sample flow evenly over the filter surface.
Have Questions?
Catalog NumberTypeGrade
FKS10528452Aerosol Monitor with PadAAWP
Catalog number FKS10528452
Price (MYR)
5,783.00
PK
Add to cart
Type:
Aerosol Monitor with Pad
Grade:
AAWP
Price (MYR)
5,783.00
PK
Add to cart
The three-part design holds the filter in-place when the top section is removed for open aerosol sampling.
The Average Background particle Count (ABC) value is marked on each monitor package.
  • Pre-assembled with filters in-place
  • Available with matched-weight membranes
Applications:
Aerosol analysis monitors are available with 0.45 μm and 0.8 μm cellulose esters membranes. Matched weight monitors contain two filters matched in weight to within 0.1 mg, eliminating test filter pre-weighing in gravimetric analysis.
Specifications
ColorWhite
ConnectorsFeMale Luer Slip
MaterialPolystyrene
Pore Size0.8 μm
Filtration Area9 cm2
For Use With (Application)Microscopic particle analysis and sample collection, environmental monitoring
GradeAAWP
Sufficient ForAsbestos, trace metals, lead, lead compounds, copper, particulate gases, sulfur dioxide, arsenic
TypeAerosol Monitor with Pad
Diameter (English)1.4 in.
Diameter (Metric)3.7 cm
Diameter (Metric) Filter3.7 cm
Unit SizePK