BLAUBRAND™ Automatic Burette with Intermediate Stopcock
BLAUBRAND™ Automatic Burette with Intermediate Stopcock
Actual product may vary
BLAUBRAND™ Automatic Burette with Intermediate Stopcock
BLAUBRAND™ Automatic Burette with Intermediate Stopcock
BLAUBRAND™ Automatic Burette with Intermediate Stopcock
Pellet pattern automatic burette includes Schellbach lateral needle-valve stopcock PTFE-key. Graduations are within the error limits defined by DIN and ISO and carry the DE-M marking.
Have Questions?
Catalog NumberCapacity (Metric)
FKS1052445250 mL
Catalog number FKS10524452
Price (MYR)
4,413.00
EA
Add to cart
Capacity (Metric):
50 mL
Price (MYR)
4,413.00
EA
Add to cart
  • Borosilicate glass construction (Boro 3.3) to DIN EN ISO 385
  • Class AS
  • DE-M marking
  • Calibrated to deliver (TD, Ex)
  • Schellbach stripe
  • Includes needle-valve stopcock with PTFE spindle (intermediate stopcock with PTFE-key).
  • Automatic zeroing.
  • Intermediate stopcock 4 NS/19 for recirculating residual liquid.
  • Total height approx. 1m with 2000mL soda-lime glass bottle (sold separately)
  • All BLAUBRAND™ automatic burettes are delivered with batch number and enclosed one batch certificate per packing unit of the manufacturer.
  • On demand also available with individual certificate, USP Individual certificate or DAkkS calibration certificate.
Specifications
Capacity (Metric)50 mL
For Use With (Equipment)Bottle, Needle-valve stopcock, PTFE spindle
MaterialBorosilicate Glass
Graduations0.1 mL
IncludesSchellbach Stripe
Stopcock MaterialPTFE Key
Tolerance±0.05 mL
TypeAutomatic Buret
ClassClass AS
Certifications/ComplianceDIN EN ISO 385, USP
Unit SizeEA