7mL Borosilicate Glass Scintillation Vial
7mL Borosilicate Glass Scintillation Vial
Actual product may vary
Fisherbrand
7mL Borosilicate Glass Scintillation Vial
Smaller capacity vial saves in solvent use and disposal
Have Questions?
Catalog NumberColor
FKS10523904White
Catalog number FKS10523904
Price (MYR)
4,023.00
PK
Add to cart
Color:
White
Price (MYR)
4,023.00
PK
Add to cart
Smaller capacity vial saves in solvent use and disposal
  • Volume: 7mL
  • Closure: White plastic screw cap with cork-backed metal foil liner
  • Body O.D.: 16.5 to 17.0 mm
  • Mouth opening: 9.4 mm minimum
Specifications
Material (Liner)Foil/Cork
Diameter (English) Outer0.66 in.
Diameter (Metric) Opening9.40 mm minimum
ColorWhite
Diameter (Metric) Outer16.5 to 17 mm
MaterialBorosilicate Glass
Volume (Metric)7 mL
Height (English)2.12 in.
Packaging TypeShrink-wrapped Tray
Volume (English)0.23 oz.
Volume (Metric) Total7 mL
Closure MaterialPlastic
Closure TypeScrew Cap
Height (Metric)54 mm
Quantity200 Cases (Vial), 200 Pk. (Cap)
TypeScintillation Vial
Unit SizePK