Transparent Polypropylene Beaker
Transparent Polypropylene Beaker
Actual product may vary
Fisherbrand
Transparent Polypropylene Beaker
Use these polypropylene beakers for a variety of general laboratory applications. VITLAB™ Transparent Polypropylene Beakers feature a stable handle easy-grip handle and an easily readable, embossed blue scale.
Have Questions?
Catalog NumberCapacity (Metric)
FKS10523822500 mL
Catalog number FKS10523822
Price (MYR)
539.00
PK
Add to cart
Capacity (Metric):
500 mL
Price (MYR)
539.00
PK
Add to cart
  • Highly transparent polypropylene beaker
  • To preserve markings, do not clean at temperatures exceeding 60°C
  • Conditionally autoclavable at 121°C (2 bar) according to DIN EN 285
  • For autoclaving, we recommend model 440941 with raised graduations
Specifications
Diameter (Metric) Outer91 mm
Capacity (Metric)500 mL
MaterialPolypropylene
Height (Metric)133 mm
Unit SizePK