Nunc™ 200mL Centrifuge Tube
Nunc™ 200mL Centrifuge Tube
Actual product may vary
Fisherbrand
Nunc™ 200mL Centrifuge Tube
Process large batches more easily with Nunc 200mL centrifuge tubes, which are made from high-purity resins and can resist forces up to 7000 RCF.
Have Questions?
Catalog NumberQuantity
FKS10519701
Catalog number FKS10519701
Price (MYR)
4,934.00
PK
Add to cart
Price (MYR)
4,934.00
PK
Add to cart
Process large batches more easily with Thermo Scientific™ Nunc™ 200mL centrifuge tubes, which are made from high-purity resins and can resist forces up to 7000 RCF.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Certifications/ComplianceMeet USP Class VI Standards
Capacity (Metric)200 mL
MaterialPolypropylene
No. per Case48
No. per Pack4
SterilitySterile
ShapeConical
Bottom ShapeConical
Closure TypePlug
Diameter (Metric)60 mm
Product TypeCentrifuge Tube
Max. RCF7000 x g
Unit SizePK