Nalgene™ Aspirator Vacuum Pump
Nalgene™ Aspirator Vacuum Pump
Actual product may vary
Nalgene™ Aspirator Vacuum Pump
Nalgene™ Aspirator Vacuum Pump
Nalgene™ Aspirator Vacuum Pump
Fisherbrand
Nalgene™ Aspirator Vacuum Pump
Attach this vacuum pump directly to sink faucets for vacuum draw. It operates efficiently with water pressures as low as 7.5 psig for minimum water consumption.
Have Questions?
Catalog NumberDescriptionMax. Vacuum
FKS10516171Vacuum Pump Aspirator28.5 in. Hg
Catalog number FKS10516171
Price (MYR)
189.00
EA
Add to cart
Description:
Vacuum Pump Aspirator
Max. Vacuum:
28.5 in. Hg
Price (MYR)
189.00
EA
Add to cart
Attach the Thermo Scientific™ Nalgene™ Aspirator Vacuum Pump directly to sink faucets for vacuum draw. It can draw 11.5L air per minute at a water flow rate of 6.5L per minute. It operates efficiently with water pressures as low as 7.5 psig for minimum water consumption and comes with an integral check valve and tubing for insertion in discharge end.
Features:
  • Attaches to a standard faucet with 3/8 in. NPT connecting thread for easy installation
  • Maximum free air pumping capacity of 11.5L per minute with a water flow rate of 6.5L per minute.
  • Operates efficiently with water pressure as low as 7.5 psig (0.517 bar) for minimum water consumption
  • Ultimate achievable vacuum of 28.5 in. (724mm) Hg
  • Discharge end equipped with 4.75 in. (12cm) length of tubing to direct discharge water toward the drain or to allow attachment to additional discharge tubing
  • Integral check valve that prevents loss of vacuum when water source is disrupted and prevents backflow
  • Polypropylene, with PE tubing
Includes:
  • Integral check valve
  • 3/8 NPT connecting thread
  • 4.75 in. (12cm) of tubing
Please recycle the corrugate cartons and LDPE bags in which your Nalgene products are packaged.
Specifications
DescriptionVacuum Pump Aspirator
No. per Pack1
No. per Case24
Max. Vacuum28.5 in. Hg
Flow Rate6.5 L/min.
TypeVacuum Pump
For Use WithFaucet
Unit SizeEA