Rubber Bellows for Automatic Burettes
Rubber Bellows for Automatic Burettes
Actual product may vary
Rubber Bellows for Automatic Burettes
With valve and tubing.
Have Questions?
Catalog NumberColor
FKS10490673Orange
Catalog number FKS10490673
Price (MYR)
247.00
EA
Add to cart
Color:
Orange
Price (MYR)
247.00
EA
Add to cart
  • Orange colored
  • Single bulb
  • Provided individually
Specifications
ColorOrange
For Use With (Equipment)Automatic Burette
MaterialRubber
No. per Pack1
IncludesTubing, Valve
TypeBellows
Unit SizeEA