Cimarec™ i Telesystem Multipoint Stirrers
Cimarec™ i Telesystem Multipoint Stirrers
Actual product may vary
Cimarec™ i Telesystem Multipoint Stirrers
Safely conduct applications with aggressive media in high-humidity environments with these multipoint stirrers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacitySpeedStirring Positions
FKS104810231500 mL/position130 to 1400 rpm6
5008800950 mL/position130 to 1400 rpm60
500880341 L/position130 to 1400 rpm15
500880771500 mL/position130 to 1400 rpm6
Catalog number FKS10481023
Price (MYR)
26,341.00
EA
Add to cart
Capacity:
1500 mL/position
Speed:
130 to 1400 rpm
Stirring Positions:
6
Price (MYR)
26,341.00
EA
Add to cart
Safely conduct applications with aggressive media in high-humidity environments with Thermo Scientific™ Cimarec™ i Telesystem Multipoint Stirrers. The stirrers are designed for integration with incubators and controlled climate chambers.
Features:
  • All models submersible for use in water baths up to 50°C
  • External controllers decrease power consumption and lower heat output
  • Chemical-resistant, sealed stainless-steel housing
  • Easy recall of last used settings
  • Operating conditions are -10°C to +56°C at 100%RH in air; 0°C to 50°C submerged in water
  • IP 68 protection; easily cleaned with running water
Effective Stirring:
  • Inductive drive ensures completely synchronized stirring at all positions
  • 6-, 15- or 60-position stirrers
  • Smooth and even stirring as low as 100rpm
  • Stir speeds up to 2000rpm
  • Stir volumes up to 2L per multipoint
Includes:
Cord set with various plugs and choice of controller: the Telemodul 20 C with microprocessor controls, adjustable power setting, and programmability or the Telemodul 40 C with higher speeds, more power settings, and PC control
Warranty:
5 years
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Speed130 to 1400 rpm
Stirring Positions6
Top Plate MaterialStainless Steel
Capacity1500 mL/position
ShapeSquare
Voltage100/240 V
Wattage5/10/15/20 w
Dimensions (L x W x H)16.5 x 9.4 x 1.4 in. (420 x 240 x 35 mm)
Height (English)1.4 in.
Height (Metric)35 mm
Length (English)16.5 in.
Length (Metric)420 mm
Width (English)9.4 in.
Width (Metric)240 mm
Certifications/ComplianceCE
DescriptionTelesystem 6 Position w/Telemodul 20 C; Capacity 1 to 1500mL per point
ModelTelesystem 6 Position w/Telemodul 20 C
Temperature Range (Metric)-10°C to +56°C (Operating)
TypeStirrer
Weight (English)16.5 lb.
Weight (Metric)7.5 kg
Unit SizeEA