Nalgene™ Unwire™ Test Tube Racks: Resmer™ Manufacturing Technology
Nalgene™ Unwire™ Test Tube Racks: Resmer™ Manufacturing Technology
Actual product may vary
Nalgene™ Unwire™ Test Tube Racks: Resmer™ Manufacturing Technology
Nalgene™ Unwire™ Test Tube Racks: Resmer™ Manufacturing Technology
Nalgene™ Unwire™ Test Tube Racks: Resmer™ Manufacturing Technology
Nalgene™ Unwire™ Test Tube Racks: Resmer™ Manufacturing Technology
Securely hold a full load of test tubes or centrifuge tubes for 13, 16, 20, 25 and 30mm diameter sizes with these strong, durable racks.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorArrayFor Use With (Equipment)
FKS10474101White4 x 1025mm Tubes
5970-0513Red6 x 1213mm Tubes
5970-0313Blue6 x 1213mm Tubes
FKS10149970Blue6 x 1213mm Tubes
5970-0413Green6 x 1213mm Tubes
FIS14-809-22White6 x 1213mm Tubes
5970-0320Blue4 x 1020mm Tubes
FKS10241861Red3 x 830mm Tubes
5970-0330Blue3 x 830mm Tubes
5970-0430Green3 x 830mm Tubes
5970-0030White3 x 830mm Tubes
Catalog number FKS10474101
Price (MYR)
314.00
EA
Add to cart
Color:
White
Array:
4 x 10
For Use With (Equipment):
25mm Tubes
Price (MYR)
314.00
EA
Add to cart
Securely hold a full load of test tubes or centrifuge tubes for 13, 16, 20, 25 and 30mm diameter sizes with Thermo Scientific™ Nalgene™ Unwire™ Test Tube Racks. Molded in a single piece for exceptional strength and durability, these racks have no coating to chip, peel or rust.
  • Smooth finish leaves fewer places for contaminants to lodge; no welds to break, no sharp edges.
  • Endplates accept large labels for easy identification of samples.
  • Higher specific gravity than water so it stays submerged in a water bath and won't float.
  • Withstands most lab chemicals and temperatures up to 121°C for a variety of lab procedures.
  • Available in six bright colors to help keep samples organized.
  • 15mL conical tubes fit 20mm racks; 50mL conical fit 30mm racks.
  • No assembly is required; rack is ready to use.
Please recycle the corrugated cartons and LDPE bags in which your Nalgene products are packaged.
Specifications
Product TypeTest Tube Rack
DescriptionTest Tube Rack Unwire™ Wht 25mm
Array4 x 10
AutoclavableAutoclavable
ColorWhite
HoldsTubes (25 mm)
MaterialRESMER
Positions40
ShapeRectangular
Shelves3 Tiers
For Use With (Equipment)25mm Tubes
Dimensions (L x W x H)11.75 x 4.75 x 3.62 in. (30 x 12.1 x 9.2 cm)
Length (Metric)30 cm
Length (English)11.75 in.
Width (Metric)12.1 cm
Width (English)4.75 in.
Height (Metric)9.2 cm
Height (English)3.62 in.
Diameter (Metric) Opening25 mm
Diameter (Metric) Inner25 mm
Unit SizeEA