Nalgene™ Disposable Polypropylene Robotic Reservoirs
Nalgene™ Disposable Polypropylene Robotic Reservoirs
Actual product may vary
Nalgene™ Disposable Polypropylene Robotic Reservoirs
Nalgene™ Disposable Polypropylene Robotic Reservoirs
Nalgene™ Disposable Polypropylene Robotic Reservoirs
Nalgene™ Disposable Polypropylene Robotic Reservoirs
Increase walk-away time and assay efficiency with these disposable polypropylene robotic reservoirs, available as a standard microplate format with 300mL volume.
Have Questions?
Catalog NumberBottom ShapeSterility
FKS10466392FlatNon-sterile
Catalog number FKS10466392
Price (MYR)
3,078.00
PK
Add to cart
Bottom Shape:
Flat
Sterility:
Non-sterile
Price (MYR)
3,078.00
PK
Add to cart
Increase walk-away time and assay efficiency with Thermo Scientific™ Nalgene™ Disposable Polypropylene Robotic Reservoirs. Featuring a standard microplate format with 300mL volume, the convoluted design minimizes dead volume and has baffles to reduce splashing. The flat bottom version is designed to meet a variety of assay needs in both manual and automated environments.
  • Convoluted bottom design is most effective with a 96-well format.
  • Flat Bottom design can be used with single, six-, 12-, 96-, 384- and 1536-well formats.
  • Polypropylene construction withstands robotic liquid handling, making it easier to run automated assays and increasing walk-away time.
  • Reservoir is compatible with most robotic arms, plate stackers (hotels), plate cranes, and platform configurations.
  • Reservoirs are RNase- & DNase-free.
  • Reservoir volume is 300mL (345mL brim capacity), with molded-in graduations every 100mL.
  • Reservoir is nestable for easy automated stacking and handling.
Warranty: 90 days
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Certifications/ComplianceMeets the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
ColorNatural
Capacity (Metric)300 mL
Dimensions (L x W x H)3.4 x 5 x 1.75 in. (86 x 128 x 43 mm)
MaterialPolypropylene
SterilityNon-sterile
TypeReservoir
Height (English)1.75 in.
Height (Metric)43 mm
Length (English)3.4 in.
Length (Metric)86 mm
No. per Case40
No. per Pack4
Width (English)5 in.
Width (Metric)128 mm
Plate CompatibilitySingle, 6-Well Plates, 12-Well Plates, 96-Well Plates, 384-Well Plates, 1536-Well Plates
Bottom ShapeFlat
Unit SizePK