Costar™ Snap Cap Microcentrifuge Tubes
Costar™ Snap Cap Microcentrifuge Tubes
Actual product may vary
Fisherbrand
Costar™ Snap Cap Microcentrifuge Tubes
Features a snap cap with a positive seal that allows for repeated opening and closing with flat-cap surfaces for convenient labeling. Corning™ Costar™ Snap Cap Microcentrifuge Tubes are compatible with leading microcentrifuge rotors.
Have Questions?
Catalog NumberColorCapacity (Metric)Packaging
FKS10429521Natural1.7 mL500/Pk.
Catalog number FKS10429521
Price (MYR)
575.00
CS
Add to cart
Color:
Natural
Capacity (Metric):
1.7 mL
Packaging:
500/Pk.
Price (MYR)
575.00
CS
Add to cart
  • Supplied nonsterile and are autoclavable.
  • External graduations and frosted writing spot for easy sample identification.
  • Positive seal design allows repeated opening and closing.
  • Flat cap surface for convenient labeling
  • Withstands a maximum RCF of 17,000 x g
  • RNase-/DNase-free
Specifications
Cap StyleSnap Cap
Certifications/ComplianceRNase, DNase-free
Closure ColorNatural
Closure MaterialPolypropylene
AutoclavableAutoclavable
Capacity (Metric)1.7 mL
ColorNatural
MaterialPolypropylene
No. per Pack500/Pk.
Packaging500/Pk.
SterilityNon-sterile
GraduatedYes
Closure TypeSnap Cap
Bottom ShapeConical
Max. RCF17,000 x g
Product TypeMicrocentrifuge Tube
Unit SizeCS