MicroAmp™ Optical 96-Well Reaction Plate
MicroAmp™ Optical 96-Well Reaction Plate
Actual product may vary
Fisherbrand
MicroAmp™ Optical 96-Well Reaction Plate
Designed to provide unmatched temperature accuracy and uniformity for fast, efficient PCR amplification
Have Questions?
Catalog NumberQuantity
FKS1041178510 Plates
Catalog number FKS10411785
Price (MYR)
742.00
EA
Add to cart
Quantity:
10 Plates
Price (MYR)
742.00
EA
Add to cart
This plate, constructed from a single rigid piece of polypropylene in a 96-well format, is compatible with Applied Biosystems 96-well Real-Time PCR systems and thermal cyclers.
  • Frosted to minimize interfering fluorescence from wells of cycling block
  • Screened to eliminate auto-fluorescent plates
  • Unique design provides barrier to ambient air to help ensure well-to-well temperature uniformity
  • Capacity: 96 x 0.2mL tubes
  • Platform: 2720 Thermal Cycler, 3130 Genetic Analyzer, 3130xl Genetic Analyzer, 3500 Dx Genetic Analyzer, 3500 Genetic Analyzer, 3500xL Dx Genetic Analyzer, 3500xL Genetic Analyzer, 3730 DNA Analyzer, 3730xl DNA Analyzer, 6100 Nucleic Acid PrepStation, 7000 System, 7300 System, 7500 System, 7900HT System, GeneAmp™ 9700, ProFlex™ PCR System, QuantStudio™, QuantStudio Dx, SimpliAmp™ Thermal Cycler, Veriti™ Dx Thermal Cycler, Veriti Thermal Cycler, ViiA™ 7 Dx System, ViiA 7 System
Applications: PCR, PCR & Real-Time PCR, Real Time PCR (qPCR)
Specifications
BarcodeNo Barcode
Certifications/ComplianceCertified DNA/RNase Free
Packaging10 Plates
Product LineMicroAmp
ColorOptical
Product TypeMicroplate
For Use With (Equipment)Thermal Cycler, Genetic Analyzer, DNA Analyzer
Skirt StyleSemi
MaterialPolypropylene
No. of Wells96
Volume (Metric) Working0.2 mL
Quantity10 Plates
Shipping ConditionRoom Temperature
Unit SizeEA
Contents & Storage
Contains ten 96-well reaction plates.
Will require either caps or full plate cover to seal (not included).
May be stored at room temperature. With proper storage, plates may be stored indefinitely.