PYREX™ Disposable Flat Bottom Screw Cap Culture Tubes, Without Caps
PYREX™ Disposable Flat Bottom Screw Cap Culture Tubes, Without Caps
Actual product may vary
PYREX™ Disposable Flat Bottom Screw Cap Culture Tubes, Without Caps
Manufactured from borosilicate glass to reduce pH changes and contaminants. Corning™ PYREX™ PYREX™ Borosilicate Glass Flat Bottom Culture Tubes are designed for both tissue culture and general bacteriological work.
Have Questions?
Catalog NumberCapacity (Metric)Length (Metric)Diameter (Metric)Thread Size
FKS1038361317 mL125 mm16 mm15-415
Catalog number FKS10383613
Price (MYR)
7,537.00
PK
Add to cart
Capacity (Metric):
17 mL
Length (Metric):
125 mm
Diameter (Metric):
16 mm
Thread Size:
15-415
Price (MYR)
7,537.00
PK
Add to cart
Tube has flat bottom and comes without marking spot. Tubes are made of borosilicate glass and are sold without caps.
For caps, see Cat. No. 99999.
Specifications
Design TypeThreaded
Capacity (Metric)17 mL
Length (Metric)125 mm
MaterialBorosilicate Glass
DisposableSingle-use
For Use WithIdeal for use with samples sensitive to leaching from Plastic tubes
Label TypeNone
PackagingBulk
Thread Size15-415
SterilityNon-sterile
AutoclavableAutoclavable
GraduatedNo
Diameter (Metric)16 mm
Closure TypeNot Included
Product TypeCulture Tube
Unit SizePK