Polypropylene Bottle Carrier
Polypropylene Bottle Carrier
Actual product may vary
Fisherbrand
Polypropylene Bottle Carrier
Carries up to six bottles with a maximum outer diameter of 95mm. Buerkle™ Polypropylene Bottle Carrier with vented sides allows easy rinsing and draining.
Have Questions?
Catalog NumberHolds
FKS10376031Bottles
Catalog number FKS10376031
Price (MYR)
232.00
EA
Add to cart
Holds:
Bottles
Price (MYR)
232.00
EA
Add to cart
  • PP
  • Suitable for food
Specifications
Product TypeBottle Carrier
HoldsBottles
MaterialPolypropylene
Positions6
Dimensions (L x W x H)310 x 210 x 305 mm
Length (Metric)310 mm
Length (English)12.2 in.
Width (Metric)210 mm
Width (English)8.26 in.
Height (Metric)305 mm
Height (English)12 in.
Unit SizeEA