Orion™ Silver/Sulphide Ion Selective Electrodes
Orion™ Silver/Sulphide Ion Selective Electrodes
Actual product may vary
Orion™ Silver/Sulphide Ion Selective Electrodes
Orion™ Silver/Sulphide Ion Selective Electrodes
Orion™ Silver/Sulphide Ion Selective Electrodes
Orion™ Silver/Sulphide Ion Selective Electrodes
Fisherbrand
Orion™ Silver/Sulphide Ion Selective Electrodes
Achieve fast, precise, and cost-effective silver and sulfide ion measurements with dual-electrode technology. Perfect for low-level cyanide and halide titrations.
Have Questions?
Catalog NumberConnector Types
FKS10374521BNC (Waterproof)
Catalog number FKS10374521
Price (MYR)
13,347.00
EA
Add to cart
Connector Types:
BNC (Waterproof)
Price (MYR)
13,347.00
EA
Add to cart
Make quick, simple, accurate and economical measurements of silver and sulfide ions in aqueous solutions with Thermo Scientific™ Orion™ Silver/Sulfide Electrodes. These electrodes have dual electrode-measuring silver and sulfide ions in aqueous solutions, and perform low-level cyanide and halide titrations.
The ThermoFisher Orion Silver/Sulfide electrode complies with SM 4500-S2- G, ASTM D4658-08 for Wastewater measurements.
Features:
  • 9616BNWP is a combination electrode with a Sure-Flow™ junction that allows easy cleaning. When conventional electrode junctions clog, the Sure-Flow junction can be opened and flushed clean simply by pressing the cap.
  • Silver/Sulfide Half-Cell Electrode 9416BN is to be used with the Orion 900200 sure flow double Junction reference electrode.
Solutions required:
  • ISA
    • 940011 ISA for silver measurements, 475 mL bottle
    • 941609 SAOB II reagent pack for sulfide measurements, 4 x 475 mL bottle
  • Electrode fill solution
    • 900002 Inner chamber filling solution for the double junction reference electrode,
    • 5 x 60 mL bottles
    • 900003 Outer chamber filling solution for the doublejunction reference electrode,
    • 5 x 60 mL bottles
  • Titrant for sulphide titration
    • 948206 0.1 M lead perchlorate solution, 475 mL bottles
Items included with:
  • 948201 Polishing strip. Qty 2.
Warranty period:
  • 12 months from date of purchase
Specifications
Connector TypesBNC (Waterproof)
DescriptionIonplus Sure-Flow solid state combination with WPBNC
Driftdrift-free potentials
JunctionSure-Flow Junction
Length (Metric) Cable1M
For Use With (Equipment)ISE Meter
Temperature (Metric) Operating0°C to 80°C
TypeCombination ISE
Unit SizeEA