Azlon™ Straight-sided Beakers
Azlon™ Straight-sided Beakers
Actual product may vary
Fisherbrand
Azlon™ Straight-sided Beakers
Manufactured from polypropylene for strength and good chemical resistance. Azlon™ Straight-sided Beakers provide an excellent solution for measuring and dispensing in a laboratory setting and are available in sizes to fit many needs.
Have Questions?
Catalog NumberCapacity (Metric)
FKS103295642000 mL
Catalog number FKS10329564
Price (MYR)
92.00
EA
Add to cart
Capacity (Metric):
2000 mL
Price (MYR)
92.00
EA
Add to cart
Manufactured from polypropylene for strength and good chemical resistance. Azlon™ Straight-sided Beakers provide an excellent solution for measuring and dispensing in a laboratory setting and are available in sizes to fit many needs.
  • Autoclavable to 121deg.C
  • Ideal for general laboratory use - straight stable shape
  • Squat form, rimmed edge, wide spout
  • Graduations are clearly printed in indelible solvent resistant ink
  • Complies with ISO 7056
Specifications
ColorTranslucent
Diameter (Metric) Outer158 mm
Diameter (Metric)Top158 mm
Capacity (Metric)2000 mL
MaterialPolypropylene
For Use With (Application)Ideal for General Laboratory Use
Height (Metric)160 mm
Increments200 mL
Quantity1/Pk.
Temperature (Metric) Max. Operating-20°C to +135°C
TypeBeaker
AutoclavableAutoclavable
GraduatedYes
Certifications/ComplianceISO 7056, FDA
Unit SizeEA