Aerosol Adapter, Luer Slip to 1/4 in.– 3/8 in. ID Hose
Aerosol Adapter, Luer Slip to 1/4 in.– 3/8 in. ID Hose
Actual product may vary
Fisherbrand
Aerosol Adapter, Luer Slip to 1/4 in.– 3/8 in. ID Hose
Adapts vacuum source to filter holders
Have Questions?
Catalog number FKS10283201
Price (MYR)
3,891.00
EA
Add to cart
Price (MYR)
3,891.00
EA
Add to cart
Millipore has a broad selection of fittings, adapters, valves, gauges, and tubing that can be used in conjunction with Millipore filtration systems. With stated exceptions, material of construction is Series 300 stainless steel.Tri-Clamp (TC) fittings are referred to by convention as 1 1/2 in., 2 in., etc. 1 1/2 in. TC fittings connect to 1 in. or 1 1/2 in. O.D. tube sizes. 2 in. TC fittings connect to 2 in. O.D. tube size only.
Specifications
Connectors0.25 to 0.37 in. I.D. Hose
For Use With (Equipment)Millipore Aerosol Analysis Monitor or Swinnex™ 47 mm Membrane Filter Holder
MaterialStainless Steel
For Use With (Application)To connect the vacuum source to any millipore aerosol analysis monitor or Swinnex™ 47 mm membrane filter holder
IncludesAerosol Adapter, Luer slip to 1/4 in.- 3/8 in. ID hose, stainless steel
TypeEnvironmental Test Kits or Supplies
Diameter (English) Inside0.25 in.
Diameter (Metric) Inside0.63 cm
Unit SizeEA