Latex Gas Sampling Balloon
Actual product may vary
Fisherbrand
Latex Gas Sampling Balloon
Gass collection bags that are constructed from latex. Kartell™ Latex Gas Sampling Balloon are best suited for collecting, containing, and carrying gas samples.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS102631835 L
FKS102037632 L
Catalog number FKS10263183
Price (MYR)
443.00
Each
Capacity (Metric):
5 L
Price (MYR)
443.00
Each
  • Cost effective
  • Ideal for collecting, containing, and carrying gas samples
Specifications
MaterialLatex
Diameter (Metric)230 mm
Product TypeGas Sampling Balloon
Capacity (Metric)5 L
Unit SizeEach