No-Wire™ Autoclavable Test Tube Racks
No-Wire™ Autoclavable Test Tube Racks
Actual product may vary
No-Wire™ Autoclavable Test Tube Racks
Submersible polypropylene test tube racks sink in water baths and maintain stability even in agitated water. Great alternative to wire racks, with price and space saving advantages over other plastic racks.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorPositions
FKS10227263Blue24
FIS22-269953Blue90
FKS10206684Blue90
Catalog number FKS10227263
Price (MYR)
433.00
EA
Add to cart
Color:
Blue
Positions:
24
Price (MYR)
433.00
EA
Add to cart
Submersible polypropylene test tube racks sink in water baths and maintain stability even in agitated water. Great alternative to wire racks, with price and space saving advantages over other plastic racks.
  • Suitable for use inside and outside the water bath to meet the demands of daily lab use
  • Heavier than water so the rack remains stable when submerged, even when agitated
  • Store flat to save space, but assembles quickly and easily
  • Features grids with numbered and lettered rows for tube identification
  • Interlocking feet facilitate easy stacking
  • Sturdy polypropylene is steam autoclavable at 121°C (250°F)
  • Round racks fit into 1000mL beaker and include removable, snap-on handle for easy retrieval
  • Rectangular racks measure 24.6 x 10.4 x 6.4cm (9.7 x 4.1 x 2.5 in.) and feature end plates for labeling
Specifications
Product TypeTube Rack
Array3 x 8
AutoclavableAutoclavable
ColorBlue
HoldsTubes (20 to 25 mm)
MaterialPolypropylene
Positions24
ShapeRectangular
Shelves3 Tiers
StackableYes
SterilityNon-sterile
Dimensions (L x W x H)9.68 x 4.09 x 2.51 in. (24.59 x 10.4 x 6.4 cm)
Length (Metric)24.59 cm
Length (English)9.68 in.
Width (Metric)10.4 cm
Width (English)4.09 in.
Height (Metric)6.4 cm
Height (English)2.51 in.
Diameter (Metric) Opening25 mm
Unit SizeEA