Fisherbrand
Eutech™ pH Electrode Storage Solution
Eutech™ offers Reference Standards and Solutions for use with Eutech™ pH electrodes
Have Questions?
Catalog NumberType
FKS10217903pH Electrode Storage Solution
Catalog number FKS10217903
Price (MYR)
221.00
Each
Type:
pH Electrode Storage Solution
Price (MYR)
221.00
Each
The measure of a liquid’s acidity and alkalinity, pH is one of the most common parameter measured in a wide variety of industries ranging from water and wastewater treatment, chemical production, agriculture research and production, environmental monitoring, chemical and life sciences research, biochemical and pharmaceutical research, electronics production to food processing and other industrial applications.
Eutech™ offers a wide range of pH buffers, calibration, and reference solutions for use with pH meters and electrodes.
Features:
  • Buffers and calibration solutions packed in 480 ml durable plastic bottles – economical and convenient for laboratory use.
Specifications
DescriptionStorage solution for pH electrodes
For Use With (Equipment)Eutech pH Electrode
TypepH Electrode Storage Solution
Unit SizeEach