Iron Wire Brushes
Iron Wire Brushes
Actual product may vary
Iron Wire Brushes
Made with natural bristles. Bochem™ Iron Wire Brushes are used for test tubes and flasks.
Have Questions?
Catalog NumberLength (Metric)Length (Metric) BrushFor Use With (Application)
FKS10204131270 mm100 mmTest tube
Catalog number FKS10204131
Price (MYR)
-
Length (Metric):
270 mm
Length (Metric) Brush:
100 mm
For Use With (Application):
Test tube
  • Iron wire, zincked
  • Natural bristles
  • For test tubes and flasks
Specifications
Product TypeTest Tube Brush
For Use With (Application)Test tube
Length (Metric)270 mm
Length (Metric) Brush100 mm
Quantity1 Ea.
Unit SizeEA