Pipette holder, vertical, polypropylene, 200mm x 75mm x 150mm
Product Image
Fisherbrand
Pipette holder, vertical, polypropylene, 200mm x 75mm x 150mm
Accomodates 8 pipettes with diameters not exceeding 7 mm, plus 8 pipettes with diameters not exceeding 15 mm. Small holes in base allow drainage.
Have Questions?
Catalog number FKS10203951
Price (MYR)
171.00
EA
Add to cart
Price (MYR)
171.00
EA
Add to cart
Accomodates 8 pipettes with diameters not exceeding 7 mm, plus 8 pipettes with diameters not exceeding 15 mm. Small holes in base allow drainage.
Specifications
HoldsPipettes with diameters not exceeding 10 mm (8), plus pipettes with diameters not exceeding 15 mm (8)
MaterialPolypropylene
Dimensions (L x W x H)200 x 75 x 150 mm
Length (Metric)200 mm
Width (Metric)75 mm
Height (Metric)150 mm
Unit SizeEA