Eye Wash Safety Stations
Eye Wash Safety Stations
Actual product may vary
Eye Wash Safety Stations
Eye wash safety stations offer built-in eye cup that allows user to be treated in any position and provides effective and immediate eye irrigation.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Height (English)Width (English)
FKS101495001000 mL15 in.17.5 in.
FIS10-028-101000 mL13.75 in.18 in.
Catalog number FKS10149500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
1000 mL
Height (English):
15 in.
Width (English):
17.5 in.
Eye wash safety stations offer built-in eye cup that allows user to be treated in any position and provides effective and immediate eye irrigation.
  • Three stations are available to suit all workstations: a single or a double station with 1000 mL (32 oz) eyewash bottle(s) or a single station with a 500 mL (16 oz) bottle
  • Shipped with empty bottles
  • Instruction diagrams on the bottle and station confirm how to use the eyewash bottle
  • Includes multi-language directions on the station (English, French, German and Spanish)
  • Single station dimensions: 44.5W x 38 cmH (17 1/2 x 15 in.) double station dimensions: 45.7W x 34.9 cmH (18 x 13.75 in.)
  • Hardware not included
Specifications
Capacity (English)33.81 oz.
Capacity (Metric)1000 mL
ColorGreen
Flow Rate1 gpm
Height (English)15 in.
Height (Metric)38.1 cm
IncludesOne empty 1000 mL Bottle, Maintenance Instructions
MaterialLDPE
MountingWall
Product TypeSingle Eyewash Station
Width (English)17.5 in.
Width (Metric)44.45 cm
Unit SizeEA