PTFE Containers
PTFE Containers
Actual product may vary
PTFE Containers
Isostatically molded from pure PTFE. Fisherbrand™ PTFE Containers contain thick wall construction.
Have Questions?
Catalog NumberCapacity (Metric)
FKS1014034260 mL
Catalog number FKS10140342
Price (MYR)
385.00
EA
Add to cart
Capacity (Metric):
60 mL
Price (MYR)
385.00
EA
Add to cart
Isostatically molded from pure PTFE. Fisherbrand™ PTFE Containers contain thick wall construction.
  • Smooth internal finish
  • Contains crew cap
  • Stackable
  • Autoclavable
Specifications
Capacity (Metric)60 mL
Capacity (English)2 oz.
ColorWhite
Diameter (Metric) Outer Neck51mm
Diameter (Metric) Inner Neck47mm
For Use With (Application)Storage and Transport of Hazardous Chemicals
IncludesClosure
Height (Metric) With Closure49mm
MaterialPTFE
Max. Temperature (Metric)280°C
Temperature Range (Metric)-200 to +280°C
Thickness (Metric)9 mm
TransparencyNo
Weight (Metric)100 g
GraduationsNo
Certifications/ComplianceCertificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
Closure IncludedYes
AutoclavableAutoclavable
GraduatedNo
Height (Metric)47 mm
Product TypeStorage Container
Closure Size60 mm
Diameter (Metric)60 mm
Closure TypeScrew Cap
Unit SizeEA