Check Order Status
Angular Beaker for use with Buerkle™ Telescopic Rod
Actual product may vary
Angular Beaker for use with Buerkle™ Telescopic Rod
The beaker is adaptable to different container shapes by adjustable angle setting (up to 90°).
Have Questions?
Catalog Number
Capacity (Metric)
FKS10140141
1000 mL
Catalog number
FKS10140141
Price (MYR)
820.00
EA
Add to cart
Capacity (Metric):
1000 mL
Price (MYR)
820.00
EA
Add to cart
Polypropylene
For industry and water treatment
Specifications
Product Type
Water Sampler
Material
Polypropylene
Autoclavable
Not Autoclavable
Capacity (English)
33.81 oz.
Capacity (Metric)
1000 mL
Style
Angular
For Use With (Application)
For Industry and Water Treatment