Nalgene™ Large Wide-Mouth Fluorinated HDPE Bottle with Closure
Nalgene™ Large Wide-Mouth Fluorinated HDPE Bottle with Closure
Actual product may vary
Nalgene™ Large Wide-Mouth Fluorinated HDPE Bottle with Closure
Nalgene™ Large Wide-Mouth Fluorinated HDPE Bottle with Closure
Nalgene™ Large Wide-Mouth Fluorinated HDPE Bottle with Closure
Fisherbrand
Nalgene™ Large Wide-Mouth Fluorinated HDPE Bottle with Closure
Prevent permeation loss of samples with the Thermo ScientificTM NalgeneTM Large Wide-Mouth Fluorinated HDPE Bottle with Closure.
Have Questions?
Catalog NumberCapacity (Metric)
FKS101266602 L
Catalog number FKS10126660
Price (MYR)
379.00
EA
Add to cart
Capacity (Metric):
2 L
Price (MYR)
379.00
EA
Add to cart
Prevent permeation loss of samples with the Thermo Scientific™ Nalgene™ Large Wide-Mouth Fluorinated HDPE Bottle with Closure. Fluorinating these high density polyethylene bottles provides a cost-effective alternative to costly fluoropolymer containers. Enhances long-term performance and prevents permeation loss.
  • Fluorinated to enhance long-term performance and prevent solvent permeation loss
  • Wide mouth allows access
  • Yields lower extractables than other bottles
  • NOTE: This bottle is not liquid leakproof
  • Note: The fluorination process leaves the bottle with a matte finish and light yellow coloration. This is normal for fluorinated bottles.
Includes: 
  • Fluorinated polypropylene screw closures
Recycling Information:
Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged.
Specifications
Capacity (Metric)2 L
ColorTranslucent
DescriptionWide-Mouth Bottle
Diameter (English)4.7 in.
Diameter (English) Inner Neck3.5 in.
Diameter (Metric) Inner Neck89 mm
MaterialFluorinated HDPE
No. per Case6
No. per Pack1
SterilityNon-sterile
Certifications/ComplianceUSP Class VI; FDA - 21 CFR
Closure IncludedYes
GraduatedNo
MouthWide
ShapeRound
Height (Metric)244 mm
Product TypeWide Mouth Bottle
Width (English)4.73 in.
Height (English)9.6 in.
Closure MaterialFluorinated Polypropylene
Closure Size100 mm
Diameter (Metric)120 mm
Closure TypeScrew Cap
Through the Closure DispensingNo
Unit SizeEA