Test Tube Baskets with Lids
Test Tube Baskets with Lids
Actual product may vary
Test Tube Baskets with Lids
Perfect for dipping, transporting and storing test tubes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDimensions (L x W x H)
FKS101141945 x 5.18 x 4 in.
FISS478499 x 9 x 9 in.
Catalog number FKS10114194
Price (MYR)
689.00
EA
Add to cart
Dimensions (L x W x H):
5 x 5.18 x 4 in.
Price (MYR)
689.00
EA
Add to cart
  • Polypropylene
  • Can be used for transporting small lab animals
Specifications
Product TypeBasket for Test Tubes
AutoclavableNot Autoclavable
ColorWhite
HoldsTest Tubes
MaterialPolypropylene
ShapeSquare
StackableYes
SterilityNon-sterile
For Use With (Equipment)Test Tubes
Size5 x 4 x 4 in.
Dimensions (L x W x H)5 x 5.18 x 4 in.
Length (Metric)12.7 cm
Length (English)5 in.
Width (Metric)13.1 cm
Width (English)5.18 in.
Height (Metric)10.1 cm
Height (English)4 in.
Unit SizeEA