Specifications
Compatible Tips0.1 to 200 mL pipettes
DescriptionSilicone adapter, 44mm L
AutoclavableAutoclavable
ColorTransparent/White
DisposableSingle-use
For Use WithMacro Pipette Controller
MaterialSilicone
Number of Channels1
SterilityNon-sterile
TypeSilicone Part
Volume (Metric)0.1 to 200 mL
For Use With (Equipment)BrandTech™ BRAND™ macro™ Pipet Controller
Length (Metric)44 mm
Unit SizeEA