Silicone Tubing
Silicone Tubing
Actual product may vary
Silicone Tubing
For consistently reliable and accurate fluid transfers.
Have Questions?
Catalog NumberDiameter (Metric) InnerThickness (Metric)
FKS101018012 mm1.0 mm
Catalog number FKS10101801
Price (MYR)
0.00
EA
Add to cart
Diameter (Metric) Inner:
2 mm
Thickness (Metric):
1.0 mm
Price (MYR)
0.00
EA
Add to cart
Available in different diameters
Specifications
Diameter (Metric) Inner2 mm
Overall Length (Metric)10 m
Thickness (Metric)1.0 mm
Unit SizeEA